Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylenediaminetetraacetic Acid Disodium Cobalt Salt Hydrate 98.0+%, TCI America™
SDP

Catalog No. E008925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E008925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E008925G Supplier TCI America Supplier No. E008925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 15137-09-4
Molecular Formula C10H14CoN2Na2O8
Molecular Weight (g/mol) 395.14
MDL Number MFCD00661153
InChI Key BVGFHSPTZBVUQN-UHFFFAOYSA-L
Synonym Cobalt Disodium Ethylenediaminetetraacetate
PubChem CID 131700210
IUPAC Name λ²-cobalt(2+) disodium hydrogen 2-[(carboxylatomethyl)({2-[(carboxylatomethyl)(hydrogen carboxylatomethyl)amino]ethyl})amino]ethanehydrogen carboxylate
SMILES [Na+].[Na+].[Co++].[O-]C(=O)CN(CCN(CC([O-])=O)CC([OH-])=O)CC([OH-])=O

Specifications

Color Blue-Purple
Quantity 25 g
Formula Weight 393.12
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Ethylenediaminetetraacetic Acid Disodium Cobalt Salt Hydrate
EINECSNumber (2)-1266
RTECSNumber GF9330000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.