accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1515-80-6

IUPAC Name: methyl (2E,4Z)-hexa-2,4-dienoate
Synonyms: methyl (2E,4Z)-hexa-2,4-dienoate
Molecular Weight (g/mol): 126.16
Molecular Formula: C7H10O2
InChi Key: KWKVAGQCDSHWFK-DNVGVPOPSA-N
SMILES: COC(=O)\C=C\C=C/C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results