Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Sorbate 98.0+%, TCI America™
SDP

Catalog No. S005625ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S005625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S005625ML Supplier TCI America Supplier No. S005625ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1515-80-6
Molecular Formula C7H10O2
Molecular Weight (g/mol) 126.16
MDL Number MFCD00038319,MFCD00038319
InChI Key KWKVAGQCDSHWFK-DNVGVPOPSA-N
Synonym Sorbic Acid Methyl Ester
PubChem CID 12221265
IUPAC Name methyl (2E,4Z)-hexa-2,4-dienoate
SMILES COC(=O)\C=C\C=C/C

Specifications

Color Yellow
Boiling Point 174°C
Quantity 25 mL
Formula Weight 126.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Sorbate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.