accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1516-08-1

IUPAC Name: (²H₅)ethan-1-(²H)ol
Synonyms: (²H₅)ethan(²H)ol
Molecular Weight (g/mol): 52.11
Molecular Formula: C2H6O
InChi Key: LFQSCWFLJHTTHZ-LIDOUZCJSA-N
SMILES: [2H]OC([2H])([2H])C([2H])([2H])[2H]
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)