accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrOSOOFFF

CAS RN 151600-02-1

IUPAC Name: 6-bromonaphthalen-2-yl trifluoromethanesulfonate
Synonyms: 6-bromonaphthalen-2-yl trifluoromethanesulfonate
Molecular Weight (g/mol): 355.13
Molecular Formula: C11H6BrF3O3S
InChi Key: FRNRXKFDZXFNKG-UHFFFAOYSA-N
SMILES: FC(F)(F)S(=O)(=O)OC1=CC=C2C=C(Br)C=CC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results