accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15163-36-7

IUPAC Name: 3-(decyldimethylazaniumyl)propane-1-sulfonate
Synonyms: capryl sultaine
Molecular Weight (g/mol): 307.49
Molecular Formula: C15H33NO3S
InChi Key: WKALLSVICJPZTM-UHFFFAOYSA-N
SMILES: CCCCCCCCCC[N+](C)(C)CCCS([O-])(=O)=O
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (3)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results