accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOHNH2HCl

CAS RN 151651-44-4

IUPAC Name: benzyl 2-amino-3-hydroxypropanoate hydrochloride
Synonyms: benzyl 2-amino-3-hydroxypropanoate hydrochloride
Molecular Weight (g/mol): 231.68
Molecular Formula: C10H14ClNO3
InChi Key: MGZWCDQAKCHOBX-UHFFFAOYNA-N
SMILES: Cl.NC(CO)C(=O)OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)