Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Serine benzyl ester hydrochloride, 95%, Thermo Scientific™

Catalog No. AAH6367806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6367806 5 g
AAH6367803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6367806 Supplier Thermo Scientific Chemicals Supplier No. H6367806

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 151651-44-4
Molecular Formula C10H14ClNO3
Molecular Weight (g/mol) 231.676
MDL Number MFCD00191017
InChI Key MGZWCDQAKCHOBX-SBSPUUFOSA-N
Synonym h-d-ser-obzl.hcl, h-d-ser-obzl hcl, d-serine benzyl ester hydrochloride, r-benzyl 2-amino-3-hydroxypropanoate hydrochloride, d-serinebenzylesterhydrochloride, benzyl 2r-2-amino-3-hydroxypropanoate hydrochloride, d-serine-obzl hcl, h-d-ser-obzlhcl, h-d-ser-obzl?cl, h-d-ser-obzl. hcl
PubChem CID 11776084
IUPAC Name benzyl (2R)-2-amino-3-hydroxypropanoate;hydrochloride
SMILES C1=CC=C(C=C1)COC(=O)C(CO)N.Cl

Specifications

Melting Point 169°C to 174°C
Quantity 5 g
Formula Weight 231.68
Percent Purity 95%
Physical Form Solid
Chemical Name or Material D-Serine benzyl ester hydrochloride
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.