accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OH

CAS RN 1517-63-1

IUPAC Name: (4-methylphenyl)(phenyl)methanol
Synonyms: phneyl-P-tolylcarbinol
Molecular Weight (g/mol): 198.27
Molecular Formula: C14H14O
InChi Key: IHASOVONMUHDND-UHFFFAOYNA-N
SMILES: CC1=CC=C(C=C1)C(O)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results