Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methylbenzhydrol 98.0+%, TCI America™
SDP

Catalog No. M117625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M117625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M117625G Supplier TCI America Supplier No. M117625G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1517-63-1
Molecular Formula C14H14O
Molecular Weight (g/mol) 198.27
MDL Number MFCD00004492
InChI Key IHASOVONMUHDND-UHFFFAOYNA-N
Synonym 4-methylbenzhydrol, phenyl p-tolyl methanol, p-methylbenzhydrol, 4-methylphenyl phenyl methanol, benzhydrol, 4-methyl, phenyl-p-tolyl-methanol, phneyl-p-tolylcarbinol, benzenemethanol, 4-methyl-.alpha.-phenyl, 4-methyldiphenylmethanol, 4-methylphenyl phenylmethan-1-ol
PubChem CID 95377
IUPAC Name (4-methylphenyl)(phenyl)methanol
SMILES CC1=CC=C(C=C1)C(O)C1=CC=CC=C1

Specifications

Melting Point 54°C
Color White-Yellow
Quantity 25 g
Formula Weight 198.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methylbenzhydrol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.