accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HO

CAS RN 1517-68-6

IUPAC Name: 1-phenylpropan-2-ol
Synonyms: 1-phenyl-2-propanol
Molecular Weight (g/mol): 136.19
Molecular Formula: C9H12O
InChi Key: WYTRYIUQUDTGSX-UHFFFAOYNA-N
SMILES: CC(O)CC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results