Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-1-Phenyl-2-propanol 96.0+%, TCI America™
SDP

Catalog No. P17801ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P17801ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P17801ML Supplier TCI America Supplier No. P17801ML

Chemical Identifiers

CAS 1517-68-6
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00066456
InChI Key WYTRYIUQUDTGSX-QMMMGPOBSA-N
PubChem CID 6994174
IUPAC Name (2S)-1-phenylpropan-2-ol
SMILES CC(CC1=CC=CC=C1)O

Specifications

Color Yellow
Boiling Point 104°C
Quantity 1 mL
Formula Weight 136.19
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material (S)-1-Phenyl-2-propanol
EINECSNumber (3)-1034
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.