accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15178-76-4

IUPAC Name: 3-[dimethyl(octyl)azaniumyl]propane-1-sulfonate
Synonyms: 3-[dimethyl(octyl)ammonio]propane-1-sulfonate
Molecular Weight (g/mol): 279.44
Molecular Formula: C13H29NO3S
InChi Key: QZRAABPTWGFNIU-UHFFFAOYSA-N
SMILES: CCCCCCCC[N+](C)(C)CCCS([O-])(=O)=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (1)
  • (1)
  • (2)
Quantity 
  • (1)
  • (3)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results