accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNNN

CAS RN 1518-16-7

IUPAC Name: 2-[4-(dicyanomethylidene)cyclohexa-2,5-dien-1-ylidene]propanedinitrile
Synonyms: tetracyanoquinodimethane tetracyano-p-quinodimethane TCNQ
Molecular Weight (g/mol): 204.19
Molecular Formula: C12H4N4
InChi Key: PCCVSPMFGIFTHU-UHFFFAOYSA-N
SMILES: N#CC(C#N)=C1C=CC(C=C1)=C(C#N)C#N
Molecular Weight (g/mol) 
  • (5)
  • (6)
Percent Purity 
  • (2)
  • (2)
  • (7)
Quantity 
  • (2)
  • (2)
  • (2)
  • (5)
Physical Form 
  • (7)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (4)
  • (5)
  • (5)
  • (6)
  • (2)
  • (2)
  • (7)
  • (2)
  • (2)
  • (2)
  • (5)
  • (7)