Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7,7,8,8-Tetracyanoquinodimethane 98.0+%, TCI America™
SDP

Catalog No. T00785G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T00785G 5 g
T007825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T00785G Supplier TCI America Supplier No. T00785G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1518-16-7
Molecular Formula C12H4N4
Molecular Weight (g/mol) 204.192
MDL Number MFCD00011664
InChI Key PCCVSPMFGIFTHU-UHFFFAOYSA-N
Synonym 7,7,8,8-tetracyanoquinodimethane, tcnq, tetracyanoquinodimethane, tetracyanoquinodimethan, 2,2'-cyclohexa-2,5-diene-1,4-diylidene dimalononitrile, tetracyano-p-quinodimethane, tetracyanoquinodimethan e, quinodimethan, tetracyano, propanedinitrile, 2,2'-2,5-cyclohexadiene-1,4-diylidene bis, 7,7,8,8-tetracyano-p-quinodimethane
PubChem CID 73697
ChEBI CHEBI:52445
IUPAC Name 2-[4-(dicyanomethylidene)cyclohexa-2,5-dien-1-ylidene]propanedinitrile
SMILES C1=CC(=C(C#N)C#N)C=CC1=C(C#N)C#N

Specifications

Quantity 5 g
UN Number 3439
Formula Weight 204.19
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 7,7,8,8-Tetracyanoquinodimethane
EINECSNumber (3)-1798
RTECSNumber GU4850000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.