CAS RN 15181-86-9
IUPAC Name:
ethyl 3-(1H-indol-3-yl)prop-2-enoate
Synonyms:
ethyl 3-(1H-indol-3-yl)prop-2-enoate
Molecular Weight (g/mol):
215.25
Molecular Formula:
C13H13NO2
InChi Key:
OQJSITNIWIYWPU-UHFFFAOYSA-N
SMILES:
CCOC(=O)C=CC1=CNC2=CC=CC=C12
1
–
1
of
1
results
1
Promotions Available
Ethyl indole-3-acrylate, 97%
Ethyl indole-3-acrylate, 97%
CAS: 15181-86-9 Molecular Formula: C13H13NO2 Molecular Weight (g/mol): 215.252 MDL Number: MFCD00051947 InChI Key: OQJSITNIWIYWPU-FPLPWBNLSA-N Synonym: ethyl 2z-3-1h-indol-3-yl prop-2-enoate PubChem CID: 13863197 IUPAC Name: ethyl (Z)-3-(1H-indol-3-yl)prop-2-enoate SMILES: CCOC(=O)C=CC1=CNC2=CC=CC=C21