accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15201-61-3

IUPAC Name: iron(3+) triperchlorate
Synonyms: iron(III) perchlorate
Molecular Weight (g/mol): 354.18
Molecular Formula: Cl3FeO12
InChi Key: LHOWRPZTCLUDOI-UHFFFAOYSA-K
SMILES: [Fe+3].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (3)
  • (2)
Grade 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results