Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Iron(III) Perchlorate Hydrate

Catalog No. 326348500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-634-8500G 500 g
32-634-8100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 32-634-8500G Supplier Sigma Aldrich Supplier No. 326348500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Useful starting material for the synthesis of a tris-salicylate Fe(III) complex having bidentate chelating units which allow two coordination modes.

Specifications

CAS 15201-61-3
Molecular Formula Cl3FeO12 -+ xH2O
Linear Formula Fe(ClO4)3 -+ xH2O
MDL Number MFCD00149705
Quantity 500 g
Synonym Perchloric acid iron(III) salt hydrate
Molecular Weight (g/mol) 354.2 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.