accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 152095-12-0

IUPAC Name: 1-{[bis(pyridin-2-yl)methylidene]amino}-3,3-dimethylthiourea
Synonyms: 1-{[bis(pyridin-2-yl)methylidene]amino}-3,3-dimethylthiourea
Molecular Weight (g/mol): 285.37
Molecular Formula: C14H15N5S
InChi Key: XOBIGRNRXCAMJQ-UHFFFAOYSA-N
SMILES: CN(C)C(=S)NN=C(C1=CC=CC=N1)C1=CC=CC=N1
Color 
  • (1)
Quantity 
  • (1)
Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results