Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dp44mT

Catalog No. AAJ64090MB
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64090MB 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ64090MB Supplier Thermo Scientific Chemicals Supplier No. J64090MB
Add to Cart

CAS: 152095-12-0 | C14H15N5S | 285.369 g/mol

An iron chelator and selective anticancer agent

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 152095-12-0
Molecular Formula C14H15N5S
Molecular Weight (g/mol) 285.369
MDL Number MFCD20527329
InChI Key XOBIGRNRXCAMJQ-UHFFFAOYSA-N
Synonym 2-(Di(pyridin-2-yl)methylene)-N,N-dimethylhydrazinecarbothioamide; Di-2-pyridylketone-4,4,-dimethyl-3-thiosemicarbazone
PubChem CID 10334137
IUPAC Name 3-(dipyridin-2-ylmethylideneamino)-1,1-dimethylthiourea
SMILES CN(C)C(=S)NN=C(C1=CC=CC=N1)C2=CC=CC=N2

Specifications

Type Dp44mT
Physical Form Powder
Color Orange to Yellow
Quantity 25 mg
Sensitivity Light sensitive
UN Number UN2811
Formula Weight 285.37
Chemical Name or Material Dp44mT
Hazard Category H301
Hazard Statement GHS H Statement
H300
Fatal if swallowed.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.