accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3H3COOHNNOOH3CH3CCH3NN(E/Z)

CAS RN 152120-54-2

IUPAC Name: tert-butyl N-({[(tert-butoxy)carbonyl]imino}(1H-pyrazol-1-yl)methyl)carbamate
Synonyms: tert-butyl N-{[(tert-butoxycarbonyl)imino](pyrazol-1-yl)methyl}carbamate
Molecular Weight (g/mol): 310.35
Molecular Formula: C14H22N4O4
InChi Key: QFNFDHNZVTWZED-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC(=NC(=O)OC(C)(C)C)N1C=CC=N1
Molecular Weight (g/mol) 
  • (3)
  • (1)
  • (5)
Quantity 
  • (3)
  • (2)
  • (4)
Percent Purity 
  • (3)
  • (2)
  • (3)
Physical Form 
  • (2)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (6)
  • (3)
  • (1)
  • (5)
  • (3)
  • (2)
  • (4)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)