Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (S)-(-)-2-Acetoxypropionyl Chloride

Catalog No. 4470565G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-705-65G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 44-705-65G Supplier Sigma Aldrich Supplier No. 4470565G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Frequently used chiral building block. Also used to resolve a bicyclic +a-hydroxylactone and to prepare chiral phosphonates used in an enantiomeric excess assay of unprotected amino acids. Useful chiral derivatizing agent.

Specifications

CAS 152120-54-2
Melting Point 86°C to 90°C
Boiling Point 202°C to 204°C
Molecular Formula C14H22N4O4
Linear Formula C14H22N4O4
Refractive Index n20/D 1.423
MDL Number MFCD01075122
Quantity 5 g
Synonym O-Acetyl-L-lactyl chloride; AP-Cl
Molecular Weight (g/mol) 310.35 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.