accessibility menu, dialog, popup

UserName

Viewing profile as user:
OONH2NNN(E/Z)

CAS RN 152120-62-2

IUPAC Name: benzyl N-[amino(1H-pyrazol-1-yl)methylidene]carbamate
Synonyms: benzyl N-[amino(pyrazol-1-yl)methylidene]carbamate
Molecular Weight (g/mol): 244.25
Molecular Formula: C12H12N4O2
InChi Key: BXUMISWXFKCEIA-UHFFFAOYSA-N
SMILES: NC(=NC(=O)OCC1=CC=CC=C1)N1C=CC=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results