Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(Carbobenzoxy)-1H-pyrazole-1-carboxamidine 98.0+%, TCI America™
SDP

Catalog No. C27095G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C27095G 5 g
C27091G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C27095G Supplier TCI America Supplier No. C27095G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 152120-62-2
Molecular Formula C12H12N4O2
Molecular Weight (g/mol) 244.254
MDL Number MFCD00274657
InChI Key BXUMISWXFKCEIA-UHFFFAOYSA-N
Synonym n-z-1h-pyrazole-1-carboxamidine, benzyl imino 1h-pyrazol-1-yl methyl carbamate, n-benzyloxycarbonyl-1h-pyrazole-1-carboxamidine, n-carbobenzoxy-1h-pyrazole-1-carboxamidine, n-z-1h-pyrazole-1-carboxamdne, benzyl n-imino 1-pyrazolyl methyl carbamate, benzyl imino-1h-pyrazol-1-yl methylcarbamate
PubChem CID 11857377
IUPAC Name benzyl (NE)-N-[amino(pyrazol-1-yl)methylidene]carbamate
SMILES C1=CC=C(C=C1)COC(=O)N=C(N)N2C=CC=N2

Specifications

Melting Point 111°C
Color White
Quantity 5 g
Formula Weight 244.25
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(Carbobenzoxy)-1H-pyrazole-1-carboxamidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.