accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOFCH3O

CAS RN 1522-41-4

IUPAC Name: ethyl 2-fluoro-3-oxobutanoate
Synonyms: ethyl 2-fluoro-3-oxobutanoate
Molecular Weight (g/mol): 148.13
Molecular Formula: C6H9FO3
InChi Key: SHTFQLHOTAJQRJ-UHFFFAOYNA-N
SMILES: CCOC(=O)C(F)C(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results