Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-Fluoroacetoacetate 95.0+%, TCI America™
SDP

Catalog No. F047125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F047125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F047125G Supplier TCI America Supplier No. F047125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1522-41-4
Molecular Formula C6H9FO3
Molecular Weight (g/mol) 148.133
MDL Number MFCD00215830
InChI Key SHTFQLHOTAJQRJ-UHFFFAOYSA-N
Synonym ethyl 2-fluoroacetoacetate, ethyl2-fluoroacetoacetate, 2-fluoro-3-oxo-butanoic acid ethyl ester, 2-fluoroacetoacetic acid ethyl ester, ethyl 2-fluoro-3-oxobutyrate, 2-fluoro-3-oxo-butyric acid ethyl ester, ethyl 2-fluoro-3-oxo-butanoate, ethyl-2-fluoroacetoacetate, ethyl-2-fluoro-3-oxobutanoate, fluoroacetoacetic acid ethyl ester
PubChem CID 230078
IUPAC Name ethyl 2-fluoro-3-oxobutanoate
SMILES CCOC(=O)C(C(=O)C)F

Specifications

Color Yellow
Boiling Point 85°C
Quantity 25 g
Formula Weight 148.13
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2-Fluoroacetoacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.