accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3C

CAS RN 15232-76-5

IUPAC Name: oct-3-yne
Synonyms: 3-octyne
Molecular Weight (g/mol): 110.2
Molecular Formula: C8H14
InChi Key: UDEISTCPVNLKRJ-UHFFFAOYSA-N
SMILES: CCCCC#CCC
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)