Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Octyne, 98%, Thermo Scientific™

Catalog No. AAA1284503
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1284503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1284503 Supplier Thermo Scientific Chemicals Supplier No. A1284503

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15232-76-5
Molecular Formula C8H14
Molecular Weight (g/mol) 110.2
MDL Number MFCD00027276
InChI Key UDEISTCPVNLKRJ-UHFFFAOYSA-N
Synonym 3-octyne, butylethylacetylene, ethylbutylacetylene, 3-octyne, homopolymer, 1-ethyl-2-butylacetylene, acmc-209d7q, c4h9c.$.cc2h5, udeistcpvnlkrj-uhfffaoysa, 3-octyne,, 3h-indolium, 2-4-methoxyphenyl methylhydrazono methyl-1,3,3-trimethyl-, formate
PubChem CID 84849
IUPAC Name oct-3-yne
SMILES CCCCC#CCC

Specifications

Melting Point -104°C
Boiling Point 133°C
Flash Point 24°C (75°F)
Odor Petroleum-like
Linear Formula CH3(CH2)3C≡CCH2CH3
Quantity 1 g
UN Number UN3295
Formula Weight 110.2
Percent Purity 98%
Chemical Name or Material 3-Octyne
Hazard Category Danger
Hazard Statement GHS H Statement
H304-H226
May be fatal if swallowed and enters airways.
Flammable liquid and vapour.
Precautionary Statement GHS P Statement
P210-P301+P330+P331-P315
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Get immediate medical advice/attention.
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.