accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3H3CH3CCFFFCFFFCFFFCFFFCu

CAS RN 152376-94-8

IUPAC Name: tetrabutylazanium tetrakis(trifluoromethanide) copper
Synonyms: tetrabutylammonium ion tetrakis(trifluoromethanide) copper
Molecular Weight (g/mol): 582.04
Molecular Formula: C20H36CuF12N
InChi Key: ONKMOBXWIMXGQS-UHFFFAOYSA-N
SMILES: [Cu].F[C-](F)F.F[C-](F)F.F[C-](F)F.F[C-](F)F.CCCC[N+](CCCC)(CCCC)CCCC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)