Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabutylammonium Tetrakis(trifluoromethyl)cuprate(III) 98.0+%, TCI America™
SDP

Catalog No. T3374200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3374200MG 200 mg
T33741G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T3374200MG Supplier TCI America Supplier No. T3374200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 152376-94-8
Molecular Formula C20H36CuF12N-3
Molecular Weight (g/mol) 582.042
InChI Key ONKMOBXWIMXGQS-UHFFFAOYSA-N
Synonym Tetrabutylammonium Tetrakis(trifluoromethyl)copper(III)
PubChem CID 124220112
IUPAC Name copper;tetrabutylazanium;trifluoromethane
SMILES CCCC[N+](CCCC)(CCCC)CCCC.[C-](F)(F)F.[C-](F)(F)F.[C-](F)(F)F.[C-](F)(F)F.[Cu]

Specifications

Melting Point 142°C
Color White
Quantity 200 mg
Formula Weight 582.04
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetrabutylammonium Tetrakis(trifluoromethyl)cuprate(III)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.