accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 152457-95-9

IUPAC Name: 2,5-bis(2,2,2-trifluoroethoxy)benzene-1-sulfonyl chloride
Synonyms: 2,5-bis(2,2,2-trifluoroethoxy)benzenesulfonyl chloride
Molecular Weight (g/mol): 372.66
Molecular Formula: C10H7ClF6O4S
InChi Key: UCHAULDYIPVTOP-UHFFFAOYSA-N
SMILES: FC(F)(F)COC1=CC(=C(OCC(F)(F)F)C=C1)S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results