Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Di(2,2,2-trifluoroethoxy)benzene-1-sulfonyl chloride, 97%, Thermo Scientific™

Catalog No. CD11334EA
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CD11334EA 10 g
CD11334DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CD11334EA Supplier Thermo Scientific Chemicals Supplier No. CD11334EA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 152457-95-9
Molecular Formula C10H7ClF6O4S
Molecular Weight (g/mol) 372.66
MDL Number MFCD00052841
InChI Key UCHAULDYIPVTOP-UHFFFAOYSA-N
Synonym 2,5-bis 2,2,2-trifluoroethoxy benzenesulfonyl chloride, 2,5-bis 2,2,2-trifluoroethoxy benzene-1-sulfonyl chloride, 2,5-bis 2,2,2-trifluoroethoxy benzenesulfonylchloride, 2,5-bis 2,2,2-trifluoroethoxy benzenesulphonyl chloride, 2,5-bis 2,2,2-trifluoroethoxy benzenesulfonyl, 2,5-bis-2,2,2-trifluoroethoxy benzenesulfonyl chloride, benzenesulfonylchloride, 2,5-bis 2,2,2-trifluoroethoxy, 2,5-bis 2,2,2-trifluoroethoxy benzenesulphonyl chloride, tech., pubchem5052, acmc-1c3ia
PubChem CID 2736077
SMILES FC(F)(F)COC1=CC(=C(OCC(F)(F)F)C=C1)S(Cl)(=O)=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 10 g
IUPAC Name 2,5-bis(2,2,2-trifluoroethoxy)benzene-1-sulfonyl chloride
Formula Weight 372.67
Percent Purity 97%
Chemical Name or Material 2,5-Di(2,2,2-trifluoroethoxy)benzene-1-sulfonyl chloride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.