CAS RN 152477-90-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
CERILLIANT™ 6-Acetylmorphine-d6 Solution, 100 μg/mL in acetonitrile, Sold by MilliporeSigma™ Supelco™
Suitable for quantitation of 6-acetylmorphine (6-MAM) levels in GC/MS or LC/MS testing for clinical toxicology, forensic analysis, urine drug testing, pain prescription monitoring and isotope dilution methods.