accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONOH3C

CAS RN 15248-39-2

IUPAC Name: 6,7-dimethoxyisoquinoline
Synonyms: 6,7-dimethoxyisoquinoline backebergine
Molecular Weight (g/mol): 189.21
Molecular Formula: C11H11NO2
InChi Key: JAJVYESKUNMYPN-UHFFFAOYSA-N
SMILES: COC1=C(OC)C=C2C=NC=CC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results