accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOOHOON(S)CH3CH3H3CCH3

CAS RN 152548-66-8

IUPAC Name: (2S)-4-(tert-butoxy)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}(methyl)amino)-4-oxobutanoic acid
Synonyms: (2S)-4-(tert-butoxy)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl](methyl)amino}-4-oxobutanoic acid
Molecular Weight (g/mol): 425.48
Molecular Formula: C24H27NO6
InChi Key: CYWWLVIEAOUXGW-FQEVSTJZSA-N
SMILES: CN([C@@H](CC(=O)OC(C)(C)C)C(O)=O)C(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)