Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fmoc-N-Me-Asp OtBu -OH, Advanced ChemTech

Catalog No. 01340591
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-340-591 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-340-591 Supplier Advanced Chem Tech Supplier No. FD21611G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 152548-66-8
Molecular Formula C24H27NO6
Molecular Weight (g/mol) 425.48
MDL Number MFCD00237027
InChI Key CYWWLVIEAOUXGW-FQEVSTJZSA-N
IUPAC Name (2S)-4-(tert-butoxy)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}(methyl)amino)-4-oxobutanoic acid
SMILES CN([C@@H](CC(=O)OC(C)(C)C)C(O)=O)C(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2

Specifications

Color White/Off-White
Quantity 1 g
Grade Biochemical
Chemical Name or Material Fmoc-N-Me-Asp OtBu -OH
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.