accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNHNOOHCH3

CAS RN 152628-03-0

IUPAC Name: 4-methyl-2-propyl-1H-1,3-benzodiazole-6-carboxylic acid
Synonyms: 7-methyl-2-propyl-3H-1,3-benzodiazole-5-carboxylic acid
Molecular Weight (g/mol): 218.26
Molecular Formula: C12H14N2O2
InChi Key: XWAJTVCEILFDGU-UHFFFAOYSA-N
SMILES: CCCC1=NC2=C(C)C=C(C=C2N1)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results