Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methyl-2-propyl-6-benzimidazolecarboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. M20005G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20005G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M20005G Supplier TCI America Supplier No. M20005G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 152628-03-0
Molecular Formula C12H14N2O2
Molecular Weight (g/mol) 218.256
MDL Number MFCD06656215
InChI Key XWAJTVCEILFDGU-UHFFFAOYSA-N
Synonym 6-Carboxy-4-methyl-2-propylbenzimidazole
PubChem CID 10262831
IUPAC Name 7-methyl-2-propyl-3H-benzimidazole-5-carboxylic acid
SMILES CCCC1=NC2=C(C=C(C=C2N1)C(=O)O)C

Specifications

Melting Point 300°C
Color White-Yellow
Quantity 5 g
Formula Weight 218.26
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methyl-2-propyl-6-benzimidazolecarboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.