accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClHGeHCH3CH3H3C

CAS RN 1529-47-1

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 155.2
Molecular Formula: C3H11ClGe
InChi Key: TWBCUFVBXMWAQL-UHFFFAOYSA-N
SMILES: Cl.C[GeH](C)C
Molecular Weight (g/mol) 
  • (4)
  • (1)
Quantity 
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (5)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (1)