Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethylgermanium chloride

Catalog No. AA3412914
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3412914 25 g
AA3412903 1 g
AA3412906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3412914 Supplier Thermo Scientific Chemicals Supplier No. 03412914
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1529-47-1 | C3H9ClGe | 153.185 g/mol

Trimethylgermanium chloride is used to prepare trimethyl(2-thienyl)germane. It is also used as alkylating and precursor to prepare Grignard reagent in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1529-47-1
Molecular Formula C3H9ClGe
Molecular Weight (g/mol) 153.185
MDL Number MFCD00000462
InChI Key ZZBNZZCHSNOXOH-UHFFFAOYSA-N
Synonym trimethylchlorogermane, trimethylgermanium chloride, chloro trimethyl germane, chlorotrimethylgermanium, trimethylgermyl chloride, ch3 3gecl, acmc-209d8g, chlorotrimethylgermane
PubChem CID 73723
IUPAC Name chloro(trimethyl)germane
SMILES C[Ge](C)(C)Cl

Specifications

Melting Point -14°C
Density 1.249 g/mL
Boiling Point 102°C
Flash Point 1°C (35°F)
Refractive Index 1.4337
Quantity 25 g
UN Number UN2924
Sensitivity Moisture sensitive
Solubility Information Immiscible with water.Miscible with ether,tetrahydrofuran,hexane,benzene,dichloromethane and chloroform.
Formula Weight 153.15
Physical Form Liquid
Chemical Name or Material Trimethylgermanium chloride
Show More Show Less
Hazard Category H225-H314
Hazard Statement GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
EINECSNumber 216-214-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.