accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClHGeH2ClHCH3H3C

CAS RN 1529-48-2

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 177.63
Molecular Formula: C2H10Cl2Ge
InChi Key: PDIYLVAYJQRPJM-UHFFFAOYSA-N
SMILES: Cl.Cl.C[GeH2]C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results