CAS RN 15291-77-7
phyproof™ Ginkgolide B, ≥98% (HPLC), MilliporeSigma™ Supelco™
This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG
Ginkgolide B from Ginkgo biloba leaves, ≥90% (HPLC), MilliporeSigma™ Supelco™
MDL Number: MFCD05662347 Synonym: BN-52021
Ginkgolide B, MilliporeSigma™ Supelco™
Ginkgolide B is a diterpenoid compound belonging to the group of ginkgolides, isolated from Ginkgo biloba leaves, exhibiting a wide spectrum of pharmacological properties and is used for the treatment of Alzheimer′s disease, cerebral insufficiency and memory loss.
Ginkgolide B
CAS: 15291-77-7 Molecular Formula: C20H24O10 Molecular Weight (g/mol): 424.402 MDL Number: MFCD05662347 InChI Key: SQOJOAFXDQDRGF-WVXYHUDCSA-N Synonym: sqojoafxdqdrgf-maggbrlhsa-n PubChem CID: 132470872 SMILES: CC1C(=O)OC2C1(C34C(=O)OC5C3(C2O)C6(C(C5)C(C)(C)C)C(C(=O)OC6O4)O)O