Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ginkgolide B, MilliporeSigma™ Supelco™

Catalog No. 111013221 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3221 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3221 Supplier MilliporeSigma Supelco Supplier No. 9497010MG
Add to Cart
Add to Cart

Ginkgolide B is a diterpenoid compound belonging to the group of ginkgolides, isolated from Ginkgo biloba leaves, exhibiting a wide spectrum of pharmacological properties and is used for the treatment of Alzheimer′s disease, cerebral insufficiency and memory loss.

Featured Industries: Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

CAS 15291-77-7
Physical Form Neat
Linear Formula C20H24O10
MDL Number MFCD05662347
Type Analytical Standard
Quantity 10 mg
Formula Weight 424.40
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.