accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1530-36-5

IUPAC Name: [(2-methylphenyl)methyl]triphenylphosphanium bromide
Synonyms: [(2-methylphenyl)methyl]triphenylphosphanium bromide
Molecular Weight (g/mol): 447.36
Molecular Formula: C26H24BrP
InChi Key: XDCQNYYELACHOT-UHFFFAOYSA-M
SMILES: [Br-].CC1=CC=CC=C1C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results