accessibility menu, dialog, popup

UserName

Viewing profile as user:
NN

CAS RN 1530-88-7

IUPAC Name: pyrrolidine-1-carbonitrile
Synonyms: pyrrolidine-1-carbonitrile
Molecular Weight (g/mol): 96.13
Molecular Formula: C5H8N2
InChi Key: QJRYYOWARFCJQZ-UHFFFAOYSA-N
SMILES: N#CN1CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results