Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyrrolidine-1-carbonitrile 98.0+%, TCI America™
SDP

Catalog No. P233725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P233725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P233725G Supplier TCI America Supplier No. P233725G

missing translation for 'validMainframeMsg'

Specifications

CAS 1530-88-7
Color Yellow
Boiling Point 110°C
Molecular Formula C5H8N2
MDL Number MFCD00003165
UN Number 3276
Quantity 25 g
Synonym 1-pyrrolidinecarbonitrile, 1-cyanopyrrolidine, n-cyanopyrrolidine, pyrrolidinecarbonitrile, cyanopyrrolidine, cyano-pyrrolidine, tetramethylenecyanamide, 1-pyrrolidine carbonitrile, d00fga
InChI Key QJRYYOWARFCJQZ-UHFFFAOYSA-N
SMILES N#CN1CCCC1
IUPAC Name pyrrolidine-1-carbonitrile
Molecular Weight (g/mol) 96.13
PubChem CID 73737
Formula Weight 96.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Pyrrolidine-1-carbonitrile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.