CAS RN 15307-78-5
CAS RN 15307-78-5
IUPAC Name:
methyl 2-{2-[(2,6-dichlorophenyl)amino]phenyl}acetate
Synonyms:
methyl 2-{2-[(2,6-dichlorophenyl)amino]phenyl}acetate
Molecular Weight (g/mol):
310.17
Molecular Formula:
C15H13Cl2NO2
InChi Key:
VETACGBDFVVKGZ-UHFFFAOYSA-N
SMILES:
COC(=O)CC1=CC=CC=C1NC1=C(Cl)C=CC=C1Cl
1
–
1
of
1
results
1
Methyl 2-(2,6-Dichloroanilino)phenylacetate 97.0+%, TCI America™

CAS: 15307-78-5 Molecular Formula: C15H13Cl2NO2 Molecular Weight (g/mol): 310.174 MDL Number: MFCD08273811 InChI Key: VETACGBDFVVKGZ-UHFFFAOYSA-N Synonym: 2-(2,6-Dichloroanilino)phenylacetic Acid Methyl Ester, Diclofenac Methyl Ester PubChem CID: 519102 IUPAC Name: methyl 2-[2-(2,6-dichloroanilino)phenyl]acetate SMILES: COC(=O)CC1=CC=CC=C1NC2=C(C=CC=C2Cl)Cl