Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-(2,6-Dichloroanilino)phenylacetate 97.0+%, TCI America™
SDP

Catalog No. M2903200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2903200MG 200 mg
M29031G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M2903200MG Supplier TCI America Supplier No. M2903200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 15307-78-5
Molecular Formula C15H13Cl2NO2
Molecular Weight (g/mol) 310.174
MDL Number MFCD08273811
InChI Key VETACGBDFVVKGZ-UHFFFAOYSA-N
Synonym 2-(2,6-Dichloroanilino)phenylacetic Acid Methyl Ester, Diclofenac Methyl Ester
PubChem CID 519102
IUPAC Name methyl 2-[2-(2,6-dichloroanilino)phenyl]acetate
SMILES COC(=O)CC1=CC=CC=C1NC2=C(C=CC=C2Cl)Cl

Specifications

Melting Point 102°C
Quantity 200 mg
Formula Weight 310.17
Percent Purity ≥97.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 2-(2,6-Dichloroanilino)phenylacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.