accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOHNClCl

CAS RN 15307-86-5

IUPAC Name: 2-{2-[(2,6-dichlorophenyl)amino]phenyl}acetic acid
Synonyms: diclofenac ProSorb-D diclofenac acid dicloreum delphimix duravolten voltaren-XR dyloject primofenac voltaren SR
Molecular Weight (g/mol): 296.15
Molecular Formula: C14H11Cl2NO2
InChi Key: DCOPUUMXTXDBNB-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC=CC=C1NC1=C(Cl)C=CC=C1Cl
Grade 
  • (1)
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results