Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2,6-Dichloroanilino)phenylacetic Acid 98.0+%, TCI America™
SDP

Catalog No. D374825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D374825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D374825G Supplier TCI America Supplier No. D374825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 15307-86-5
Molecular Formula C14H11Cl2NO2
Molecular Weight (g/mol) 296.15
MDL Number MFCD00056694
InChI Key DCOPUUMXTXDBNB-UHFFFAOYSA-N
Synonym Diclofenac
PubChem CID 3033
ChEBI CHEBI:47381
IUPAC Name 2-{2-[(2,6-dichlorophenyl)amino]phenyl}acetic acid
SMILES OC(=O)CC1=CC=CC=C1NC1=C(Cl)C=CC=C1Cl

Specifications

Melting Point 158°C
Color White-Yellow
UN Number 2811
Quantity 25 g
Formula Weight 296.15
Percent Purity ≥98.0% (GC,T)
Grade Reagent
Physical Form Crystalline Powder
Chemical Name or Material 2-(2,6-Dichloroanilino)phenylacetic Acid
RTECSNumber AG6310000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.